Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-12 p80 Recombinant Protein, PeproTech®

Catalog No. 502722981
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2.5 x 100 μg
5 x 100 μg
50 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-272-2981 2.5 x 100 μg
20012P80H1 100 μg
20012P80H10 10 μg
50-272-2980 5 x 100 μg
20012P80H2 2 μg
20012P80H50 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 50-272-2981 Supplier Gibco™ Supplier No. 20012P80H250
Add to Cart
Add to Cart

Recombinant Protein

200-12P80H-1MG will be provided as 10 x 100 μg (200-12P80H-100UG). 200-12P80H-250UG will be provided as multiple 50 μg (200-12P80H-50UG) and 100 μg (200-12P80H-100UG) vials. 200-12P80H-500UG will be provided as 5 x 100 μg (200-12P80H-100UG). Recombinant Human IL-12 p80 is an 80.0 kDa disulfide linked homodimer consisting of two p40 chains of IL-12. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-12 is a disulfide-linked heterodimeric protein (p70), composed of two subunits, p35 and p40, which are encoded by two different genes. Accumulating data indicates that p40 secretion precedes that of IL-12 expression. In addition, to its ability to covalently bind to p35 to form IL-12, p40 can bind to p19 to form IL-23, or it can form the homodimer designated IL-12 p80. Elevated levels of IL-12 p80 correlate to macrophage recruitment and increased inflammation in asthma and respiratory viral infection models.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29460
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3593
Molecular Weight (g/mol) 80 kDa
Name Human IL-12 p80
Quantity 2.5 x 100 μg
Source Insect cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias IL-12; Interleukin
Common Name IL-12 p80
Gene Symbol IL12B
Conjugate Unconjugated
Recombinant Recombinant
Sequence p40 subunit: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS
Content And Storage -20°C
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.