Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-12 R beta 1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Catalog No. p-4715044
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB839N025 25 Tests
FAB839N100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB839N025 Supplier R&D Systems Supplier No. FAB839N025
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

IL-12 R beta 1 Monoclonal specifically detects IL-12 R beta 1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-12 R beta 1
Applications Flow Cytometry
Classification Monoclonal
Clone 69310
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P42701
Gene Alias CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL12R beta 1, IL-12R subunit beta-1, IL12RB, IL12RB1, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454
Gene Symbols IL12RB1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 1 Cys24-Glu540 Accession # P42701
Purification Method Protein A or G purified from ascites
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3594
Test Specificity Detects human IL-12 R beta 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-12 R beta 1 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.