Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-13 R alpha 2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-5379948
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6141SP 25 μg
MAB6141100 100 μg
MAB6141500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB6141SP Supplier R&D Systems Supplier No. MAB6141SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

IL-13R alpha 2 Monoclonal specifically detects IL-13R alpha 2 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-13R alpha 2
Applications ELISA
Classification Monoclonal
Clone 83807
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q14627
Gene Alias cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL13R alpha 2, IL-13R subunit alpha-2, IL13RA2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein
Gene Symbols IL13RA2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-13 R alpha 2 Cys22-Leu342 Accession # Q14627
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3598
Test Specificity Detects human IL-13 R alpha 2 in ELISAs. In sandwich ELISAs, no cross-reactivity or interference was observed with recombinant human (rh) IL-4 R, rhIL-5 R alpha , rhIL-9 R, rhIL-13, rhIL-13 R alpha 1, recombinant mouse (rm) IL-13, rmIL-13 R alpha 1, or rmIL-13 R alpha 2.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.