Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-13 Recombinant Protein, PeproTech®

Catalog No. 2001310UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
200-13-10UG 10 μg
20013100UG 100 μg
200-13-1MG 1 mg
20013250UG 250 μg
200-13-2UG 2 μg
20013500UG 500 μg
200-13-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 200-13-10UG Supplier Gibco™ Supplier No. 2001310UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-13 is a 12.6 kDa protein consisting of 115 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35225
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 3596
Molecular Weight (g/mol) 12.6 kDa
Name Human IL-13
Quantity 10 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Common Name IL-13
Gene Symbol Il13
Biological Activity The ED50 as determined by the dose-dependent proliferation of TF-1 cells was ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence MSPGPVPPST ALRELIEELV NITQNQKAPL CNGSMVWSIN LTAGMYCAAL ESLINVSGCS AIEKTQRMLS GFCPHKVSAG QFSSLHVRDT KIEVAQFVKD LLLHLKKLFR EGRFN
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.