Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-13 variant Recombinant Protein, PeproTech®

Catalog No. 20013A500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20013A500UG 500 μg
20013A100UG 100 μg
20013A10UG 10 μg
200-13A-1MG 1 mg
20013A250UG 250 μg
200-13A-2UG 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 20013A500UG Supplier Gibco™ Supplier No. 20013A500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's genetic variant, termed Recombinant Human IL-13 Variant, is a mature 115 amino acid protein with a substitution of Q for R at position 113. The calculated molecular weight of Recombinant Human IL-13 Variant is 12.6 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and also by mast cells and NK cells. Targeted deletion of IL-13 in mice resulted in impaired Th2 cell development and indicated an important role for IL-13 in the expulsion of gastrointestinal parasites. IL-13 exerts anti-inflammatory effects on monocytes and macrophages and it inhibits the expression of inflammatory cytokines such as IL-1beta, TNF-alpha, IL-6 and IL-8. IL-13 has also been shown to enhance B cell proliferation and to induce isotype switching, resulting in increased production of IgE. Blocking of IL-13 activity inhibits the pathophysiology of asthma. Human and murine IL-13 are cross-species reactive. A variant of IL-13 shows enhanced functional activity compared with the wild type IL-13.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35225
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3596
Molecular Weight (g/mol) 12.6 kDa
Name Human IL-13 variant
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias IL-13; Interleukin
Common Name IL-13 variant
Gene Symbol IL13
Biological Activity This IL-13 analog shows a two fold increase, relative to wild type IL-13, in bioactivity as measured by the in vitro dose dependent activation of STAT6 and IL-13 dependent gene induction in transfected A201.1 cells. This analog has also been shown to exhibit increased in vivo activity compared to wild type IL-13, as measured by the induction of airway hyper-responsiveness.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MSPGPVPPST ALRELIEELV NITQNQKAPL CNGSMVWSIN LTAGMYCAAL ESLINVSGCS AIEKTQRMLS GFCPHKVSAG QFSSLHVRDT KIEVAQFVKD LLLHLKKLFR EGQFN
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.