Learn More
PBL Assay Science VeriKine-HS Human Interleukin-15 (IL-15) ELISA, Plasma/Serum/Tissue Culture Media (TCM) (High Sensitivity) [1x96 well plate]
Supplier: PBL Assay Science 417021
VeriKine-HS Human Interleukin-15 ELISA is manufactured by PBL Assay Science (Catalog #41702-1) for research use only (RUO) and packaged as a 1x96-well plate.This kit is Serum, Plasma, and Tissue Culture Media (TCM) compatible, quantifies Human IL-15, and has an assay range of 0.51-32.8 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.