Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-15RA ELISA Kit

Catalog No. EH258RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH258RB 96 Tests
EH258RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH258RB Supplier Invitrogen™ Supplier No. EH258RB
Add to Cart
Add to Cart

ELISA

Human IL-15RA quantitates human IL-15RA in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-15RA.

Interleukin 15 (IL-15RA) is a cytokine receptor that specifically binds IL-15 with high affinity. IL-15 regulates T and natural killer cell activation and proliferation. The IL-15 and IL-2 receptors share two subunits, IL-2R beta and IL-2R gamma, and IL-15RA is structurally related to IL-2RA, an additional IL-2-specific alpha subunit necessary for high affinity IL-2 binding. Unlike IL-2RA, IL-15RA is capable of binding IL-15 with high affinity independent of the other subunits. IL-15RA is thought to enhance cell proliferation and expression of apoptosis inhibitor Bcl-xL and Bcl-2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AA690181; CD215; IL 15 receptor; IL 15 receptor subunit a; IL 15 R; IL 15 R alpha; IL 15 R a; IL15 receptor; IL15 receptor subunit alpha; IL-15 receptor subunit alpha; IL15 receptor subunit a; IL15R; IL15R alpha; IL15R a; Il15ra; IL-15 RA; IL-15 R-alpha; Interleukin 15 receptor; interleukin 15 receptor alpha isoform EM2; interleukin 15 receptor alpha isoform IC2; interleukin 15 receptor alpha isoform IC3; interleukin 15 receptor alpha isoform IC4; interleukin 15 receptor alpha isoform IC5; interleukin 15 receptor alpha isoform IC6; interleukin 15 receptor alpha isoform IC7; interleukin 15 receptor alpha isoform IC8; interleukin 15 receptor subunit alpha; Interleukin 15 receptor subunit a; interleukin 15 receptor, alpha; interleukin 15 receptor, alpha chain; interleukin-15 receptor subunit alpha; sIL 15 R; sIL-15 receptor subunit alpha; sIL15R; sIL-15 RA; sIL-15 R-alpha; soluble IL 15 R; soluble IL15R; Soluble interleukin-15 receptor subunit alpha
Assay Range 0.08 to 20 ng/mL
Assay Sensitivity 0.08 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3601
Gene Symbol IL15RA
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target IL15RA
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.