Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-16 Recombinant Protein

Catalog No. PIRP8609
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP8609 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP8609 Supplier Invitrogen™ Supplier No. RP8609

Recombinant Protein

IL-16 (human) Recombinant Protein, RP-8609, from Invitrogen.

Interleukin 16 (IL-16) is produced primarily by CD4+ and CD8+ T cells and acts as a chemo-attractant for lymphocytes, monocytes, eosinophils, dendritic cells and Langerhans cells.  Additionally, IL-16 has been reported to upregulate IL-2 receptor (CD25), induce progression of cells to the G1 phase and suppress HIV & SIV replication.   Recombinant human IL-16 is a non-glycosylated protein, containing130 amino acids, with a molecular weight of 13.5 kDa. Purity is typically greater than 95% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the ability to chemoattract human T lymphocytes using a concentration range of 10-100 ng/mL. The specific activity of this protein is 1x10^5 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: MPDLNSSTDS AASASAASDV SVESTAEATV CTVTLEKMSA GLGFSLEGGK GSLHGDKPLT INRIFKGAAS EQSETVQPGD EILQLGGTAM QGLTRFEAWN IIKALPDGPV TIVIRRKSLQ SKETTAAGDS.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14005
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 10mM sodium phosphate with no preservative; pH 7.5
Gene ID (Entrez) 3603
Molecular Weight (g/mol) ∽13.5
Name Human IL-16
pH Range 7.5
Quantity 10 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias H-IL-16
Common Name Human IL-16
Gene Symbol IL16
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitro research only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.