Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-17 Quantikine HS ELISA Kit
SDP

Catalog No. HS170 Shop All Novus Products
Change view
Click to view available options
Content And Storage:
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
HS170 Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. HS170 Supplier R&D Systems™ Supplier No. HS170

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human IL-17 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-17 Quantikine HS ELISA Kit quantifies human IL-17/IL-17A in serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Type Human IL-17 Quantikine HS ELISA Kit
Gene ID (Entrez) 3605 (Human); 16171 (Mouse); 301289 (Rat); 481837 (Canine)
Includes Microplate, conjugate, standards, assay diluents, calibrator diluents, wash buffer, color reagent, stop Solution, plate sealers
Intended Use Measure IL-17 levels in serum and plasma
Product Line R&D Systems
Time Range 4 hours

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.