Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-17A, Biotinylated, Carrier-Free Recombinant Protein

Catalog No. BT7955025CF
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT7955025CF 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT7955025CF Supplier R&D Systems™ Supplier No. BT7955025/CF
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-17A, Biotinylated Protein is derived from HEK293. The Recombinant Human IL-17A, Biotinylated Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q16552
For Use With (Application) Bioactivity
Gene ID (Entrez) 3605
Molecular Weight (g/mol) Observed M.W.: 14-23kDa, reducing conditions, Theoretical M.W.: 15kDa (unlabeled)
Quantity 25 μg
Source Human embryonic kidney cell,HEK293-derived human IL-17/IL-17A protein Gly24-Ala155
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Common Name IL-17/IL-17A
Gene Symbol IL17A
Biological Activity Measured by its ability to induce IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells. Yao, Z. et al. (1995) Immunity 3:811. The ED50 for this effect is typically 1.5-7.5ng/mL.
Conjugate Biotin
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Gly24-Ala155
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.