Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-17B Recombinant Protein, PeproTech®

Catalog No. 2002801M
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-028-01M 2 x 500 μg
20028100UG 100 μg
20028250UG 250 μg
200-28-25UG 25 μg
20028500UG 500 μg
200-28-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 20-028-01M Supplier Gibco™ Supplier No. 2002801M
Add to Cart
Add to Cart

Recombinant Protein

200-28-1MG will be provided as 2 x 500 μg (200-28-500UG). Recombinant Human IL-17B is a 36.6 kDa non-disulfide-linked homodimer of two 161 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a T cell-derived cytokine that shares sequence similarity with IL17. This cytokine was reported to stimulate the release of TNF alpha (TNF) and IL1 beta (IL1B) from a monocytic cell line. Immunohistochemical analysis of several nerve tissues indicated that this cytokine is primarily localized to neuronal cell bodies.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UHF5
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 27190
Molecular Weight (g/mol) 36.6 kDa
Name Human IL-17 B
Quantity 2 x 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 1110006O16Rik; 1700006N07Rik; Cytokine C x 1; Cytokine Zcyto7; Cytokine-like protein ZCYTO7; gene for cytokine-like protein; IL17B; IL-17 B; IL20; IL-20; Interleukin; interleukin 17 B; interleukin 20; interleukin-17 beta; interleukin-17 B; Interleukin20; Interleukin-20; neuronal interleukin-17 related factor; neuronal interleukin-17-related factor; NIRF; UNQ516/PRO1031; ZCYTO7
Common Name IL17B
Gene Symbol IL17B
Biological Activity Determined by its ability to induce IL-8 in. human PBMCs using a concentration range of 10 ng-100 ng. Note: Results may vary with PBMC donors.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQPRSPKSKR KGQGRPGPLA PGPHQVPLDL VSRMKPYARM EEYERNIEEM VAQLRNSSEL AQRKCEVNLQ LWMSNKRSLS PWGYSINHDP SRIPVDLPEA RCLCLGCVNP FTMQEDRSMV SVPVFSQVPV RRRLCPPPPR TGPCRQRAVM ETIAVGCTCI F
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.