Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-17D, Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF2002725UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2002725UG 25 μg
AF20027100 100 μg
AF200271MG 1 mg
AF20027500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AF2002725UG Supplier Gibco™ Supplier No. AF2002725UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-17D is a 40.5 kDa disulfide-linked homodimer of two 185 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. The treatment of endothelial cells with this cytokine has been shown to stimulate the production of other cytokines including IL6, IL8 and CSF2/ GM-CSF. The increased expression of IL8 induced by this cytokine was found to be NF-kappa B-dependent.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8TAD2
For Use With (Application) ELISA, Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 53342
Molecular Weight (g/mol) 40.5 kDa
Name Human IL-17 D, Animal-Free
Quantity 25 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias AI462269; FLJ30846; IL17D; IL-17 D; IL-22; IL-27; Il27a; IL-3 R; Interleukin; interleukin 17 D; interleukin 27 A; interleukin-17 D; Interleukin-27; UNQ3096/PRO21175
Common Name IL17D
Gene Symbol IL17D
Biological Activity Measured by its binding ability in a functional ELISA.
Conjugate Unconjugated
Recombinant Recombinant
Sequence APRAGRRPAR PRGCADRPEE LLEQLYGRLA AGVLSAFHHT LQLGPREQAR NASCPAGGRP ADRRFRPPTN LRSVSPWAYR ISYDPARYPR YLPEAYCLCR GCLTGLFGEE DVRFRSAPVY MPTVVLRRTP ACAGGRSVYT EAYVTIPVGC TCVPEPEKDA DSINSSIDKQ GAKLLLGPND APAGP
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.