Learn More
ACROBiosystems Biotinylated Human IL-17E / IL-25 Protein, His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems IL5H82E125UG
Encompass_Preferred
Biotinylated Human IL-17E Protein, His,Avitag (IL5-H82E1) is expressed from human 293 cells (HEK293). It contains AA Tyr 33 - Gly 177 (Accession # NP_073626.1).
Catalog No. 50-233-1751