Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-17E (IL-25) Recombinant Protein, PeproTech®

Catalog No. 20024100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
25 μg
5 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20024100UG 100 μg
20-024-01M 2 x 500 μg
200-24-25UG 25 μg
20024500UG 500 μg
200-24-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 20024100UG Supplier Gibco™ Supplier No. 20024100UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

200-24-1MG will be provided as 2 x 500 μg (200-24-500UG). Recombinant Human IL-17E is a 33.8 kDa disulfide-linked homodimer of two 146 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H293
For Use With (Application) Control, Western Blot Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 64806
Molecular Weight (g/mol) 33.8 kDa
Name Human IL-17E (IL-25)
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias D17Wsu104e; il 17e; Il17e; IL-17E; Il25; IL-25; ILN; Interleukin; interleukin 17E; interleukin 25; interleukin-17E; Interleukin-25; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; RGD1561632; UNQ3120/PRO10272; UPF0556 protein C19orf10 homolog
Common Name IL-25
Gene Symbol IL25
Biological Activity Determined by its ability to induce IL-8 in. human PBMCs using a concentration range of 10-100 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MYSHWPSCCP SKGQDTSEEL LRWSTVPVPP LEPARPNRHP ESCRASEDGP LNSRAISPWR YELDRDLNRL PQDLYHARCL CPHCVSLQTG SHMDPRGNSE LLYHNQTVFY RRPCHGEKGT HKGYCLERRL YRVSLACVCV RPRVMG
Content And Storage -20°C
Activity Determined by its ability to induce IL-8 in human PBMCs using a concentration range of 10-100 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.