Learn More
FUJIFILM IRVINE SCIENTIFIC INC Human IL-17E (IL-25) , Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 100149100UG
Encompass_Preferred
Interleukin 17E (IL-17E), also known as IL-25, is a member of the IL-17 family of cytokines. IL-17E binds to the IL-17RB receptor to stimulate the secretion of the proinflammatory interleukin 8 (IL-8) chemokine and to induce the activation of nuclear factor kappa-light-chain-enhancer of activated B cells (NF-KB).
Catalog No. 50-900-503
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.