Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-17RA/IL-17R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7064611
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1771SP 25 μg
MAB1771 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1771SP Supplier R&D Systems Supplier No. MAB1771SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17RA/IL-17R Monoclonal specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-17RA/IL-17R
Applications Western Blot, ELISA
Classification Monoclonal
Clone 133621
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96F46
Gene Alias CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
Gene Symbols IL17RA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-17 RA/IL-17 R Leu33-Trp320 Accession # Q96F46
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23765
Test Specificity Detects human IL-17 RA/IL-17 R in ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.