Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-18BP Fc Recombinant Protein, PeproTech®

Catalog No. 20018BP1MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20018BP1MG 1 mg
20018BP100 100 μg
20018BP20UG 20 μg
20018BP250 250 μg
20018BP500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 20018BP1MG Supplier Gibco™ Supplier No. 20018BP1MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human IL-18BP Fc is a glycosylated, disulfide-linked homodimer of 792 amino-acid-residues whose monomer consists of a 163-amino-acid portion of secreted IL-18BP fused to the 231-amino-acid length Fc portion of human IgG by two glycine residues. The calculated molecular weight of monomeric CHO cell-derived Recombinant Human IL-18BP Fc is 43.6 kDa; however, due to glycosylation, it migrates at an apparent molecular weight of approximately 60-65 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin-18 binding protein (IL-18BP) is a constitutively secreted glycoprotein that acts as a natural antagonist for interleukin-18 (IL-18) by preventing interaction between IL-18 and its receptors. Through this binding, IL-18BP neutralizes the proinflammatory role of IL-18 in cell-mediated immune responses, resulting in reduced IFN-gamma production by T helper type I cells and inhibition of allospecific cytotoxic T lymphocyte (CTL) activity through augmented natural killer (NK) cell cytotoxicity. As an inhibitor of IL-18 and an essential component of skin homeostasis, IL-18BP has been linked to a variety of diseases, including eczema, asthma, and the autoimmune condition, Crohn's disease. The upregulation of IL-18BP by keratinocytes expressing the human papillomavirus (HPV) E7 oncoprotein has been shown to decrease IL-18-induced IFN-gamma production and IL-18-mediated T cell activation. Elevated levels of IL-18 and IL-18BP, as well as proinflammatory cytokines IFN-gamma, TNF-alpha, IL-1beta and IL-8, have been linked to chronic inflammation and observed in macrophages (including Kupffer cells) isolated from lesions and intestinal tissues of Crohn's disease patients. As a member of the immunoglobulin-like class of receptors, IL-18BP contains a single immunoglobulin (Ig) domain; although two (b and d) of the four identified human isoforms (a-d) contain incomplete domains that greatly reduce their binding affinity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95998
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 10068
Molecular Weight (g/mol) 87.2 kDa
Name Human IL-18 BP Fc
Quantity 1 mg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Igifbp; Il18bp; IL-18 BP; IL18BPa; interferon gamma inducing factor binding protein; interferon gamma-inducing factor-binding protein; interleukin 18 binding protein; interleukin-18 binding protein; interleukin-18-binding protein; MC51L-53 L-54 L homolog gene product; MC54L; Tadekinig-alfa
Common Name IL18BP
Gene Symbol IL18BP
Biological Activity Determined by its ability to neutralize IL-18 induced IFN-gamma production in KG-1 cells.
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence TPVSQTTTAA TASVRSTKDP CPSQPPVFPA AKQCPALEVT WPEVEVPLNG TLSLSCVACS RFPNFSILYW LGNGSFIEHL PGRLWEGSTS RERGSTGTQL CKALVLEQLT PALHSTNFSC VLVDPEQVVQ RHVVLAQLWA GLRATLPPTQ EALPSSHSSP QQQGGPKSCD KTHTCPPCPA PELLGGPSVF LFPPKPKDTL MISRTPEVTC VVVDVSHEDP EVKFNWYVDG VEVHNAKTKP REEQYNSTYR VVSVLTVLHQ DWLNGKEYKC KVSNKALPAP IEKTISKAKG QPREPQVYTL PPSRDELTKN QVSLTCLVKG FYPSDIAVEW ESNGQPENNY KTTPPVLDSD GSFFLYSKLT VDKSRWQQGN VFSCSVMHEA LHNHYTQKSL SLSPGK
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.