Learn More
Invitrogen™ Human IL-1F5 ELISA Kit
Description
Human IL-1F5 quantitates human IL-1F5 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-1F5.
Specifications
Specifications
| Accession Number | AI413231; FIL1; FIL1 delta; FIL1(DELTA); FIL1D; Fil1delta; IL-1 delta; IL-1 related protein 3; Il1f5; IL-1F5; IL1F5 (Canonical product IL-1F5a); IL-1F5 (IL-1 HY1, FIL1-delta, IL-1 RP3, IL-1L1, IL-1-delta); Il1h3; Il-1h3; IL1HY1; IL-1 HY1; IL1L1; IL-1L1; IL-1 ra homolog 1; IL-1-related protein 3; IL1RP3; IL-1 RP3; IL36RA; IL-36 Ra; IL36RN; interleukin 1 family, member 5 (delta); interleukin 1 receptor antagonist homolog 1; interleukin 36 receptor antagonist; Interleukin-1 delta; interleukin-1 family member 5; Interleukin-1 homolog 3; interleukin-1 HY1; Interleukin-1 receptor antagonist homolog 1; interleukin-1-like protein 1; Interleukin-36 receptor antagonist protein; MGC29840; PSORP; PSORS14; UNQ1896/PRO4342 |
| Assay Range | 0.07 to 50 ng/mL |
| Assay Sensitivity | 0.07 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 26525 |
| Gene Symbol | IL36RN |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.