Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-1RA, Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF20001RA1
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF20001RA1 1 mg
AF20001RA 500 μg
AF20001RA20 20 μg
20-001-RA100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AF20001RA1 Supplier Gibco™ Supplier No. AF20001RA1MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-1RA is a 17.2 kDa protein consisting of 153 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18510
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3557
Molecular Weight (g/mol) 17.2 kDa
Name Human IL-1 RA, Animal-Free
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias Anakinra; CD121A; cytokine; D2S1473; DIRA; F630041P17Rik; ICIL-1 RA; IL 1 receptor antagonist; IL1 inhibitor; il1 receptor; il-1 receptor; IL1 receptor antagonist; IL1F3; IL1R; il-1 r; IL1RA; IL-1 RA; IL1-RA; IL-1 ra3; IL-1 R-alpha; IL1RN; IL-1 RN; IL1RN (IL1F3); interleukin 1 receptor antagonist; interleukin-1 receptor; interleukin-1 receptor antagonist; interleukin-1 receptor antagonist protein; interleukin-1 receptor antagonist secretory form; intracellular IL-1 receptor antagonist type II; intracellular interleukin-1 receptor antagonist (icIL-1 ra); IRAP; IRAP1; MGC10430; MVCD4; P80; type II interleukin-1 receptor antagonist
Common Name IL1RA
Gene Symbol IL1RN
Biological Activity Determined by its ability to inhibit the IL-1 alpha stimulation of murine D10S cell. The expected ED50 is 20-40 ng/ml in the presence of 50 pg/ml of IL-1 alpha.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MRPSGRKSSK MQAFRIWDVN QKTFYLRNNQ LVAGYLQGPN VNLEEKIDVV PIEPHALFLG IHGGKMCLSC VKSGDETRLQ LEAVNITDLS ENRKQDKRFA FIRSDSGPTT SFESAACPGW FLCTAMEADQ PVSLTNMPDE GVMVTKFYFQ EDE
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.