Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-1RA Recombinant Protein

Catalog No. PIRP10925
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP10925 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP10925 Supplier Invitrogen™ Supplier No. RP10925
Add to Cart
Add to Cart

Recombinant Protein

The RP-10925 protein is a recombinant protein suitable for use as a control in ELISA and Western blot applications. Reconstitute with sterile, distilled water no less than 100 μg/mL.

The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18510
For Use With (Application) Control
Formulation protein with no preservative
Gene ID (Entrez) 3557
Molecular Weight (g/mol) 17 kDa
Name Human IL-1 RA
Quantity 20 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias Anakinra; CD121A; cytokine; D2S1473; DIRA; F630041P17Rik; ICIL-1 RA; IL 1 receptor antagonist; IL1 inhibitor; il1 receptor; il-1 receptor; IL1 receptor antagonist; IL1F3; IL1R; il-1 r; IL1RA; IL-1 RA; IL1-RA; IL-1 ra3; IL-1 R-alpha; IL1RN; IL-1 RN; IL1RN (IL1F3); interleukin 1 receptor antagonist; interleukin-1 receptor; interleukin-1 receptor antagonist; interleukin-1 receptor antagonist protein; interleukin-1 receptor antagonist secretory form; intracellular IL-1 receptor antagonist type II; intracellular interleukin-1 receptor antagonist (icIL-1 ra); IRAP; IRAP1; MGC10430; MVCD4; P80; type II interleukin-1 receptor antagonist
Common Name IL1RA
Gene Symbol IL1RN
Biological Activity ED50 = 0.5 ng/mL; determined by the dose-dependent inhibition of D10S cells stimulated with IL-1.
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Sequence Human IL-1 RA recombinant protein contains 153 amino acids
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Lyophilized
Protein Subtype Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.