Learn More
Novus Biologicals™ Human IL-2 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP189875
Description
The Human IL-2 ELISA Kit (Colorimetric) measures human IL-2 in cell culture supernatant, serum, plasma (edta, citrate, heparin), amniotic fluid, urine, white blood.Specifications
18.8 - 1,200pg/mL | |
ELISA Kit | |
ELISA | |
IL2 | |
96 Tests | |
Storage of components varies. See protocol for specific instructions. | |
IL-2 |
9.9pg/mL | |
Amniotic Fluid, Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine, White Blood Cells | |
3558 | |
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-2, 1 vial (100 ul) Biotin-Conjugate anti-human IL-2 polyclonal antibody, Streptavidin-HRP, Human IL-2 Standard lyophilized, 2.4ng/mL upon reconstitution, Control high, lyophilized, Control low, lyophilized, Sample Diluent, Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films | |
Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity | |
aldesleukin, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF |