Learn More
Invitrogen™ Human IL-2 ELISA Standard 10X Recombinant Protein
Description
Description: Human IL-2 (also known as TCGF) is a ~15 kDa factor produced mainly by activated CD4+ T cells. IL-2 induces cell cycle progression of resting cells in an antigen non-specific manner and allows clonal expansion of activated T cells. IL-2 also acts on activated B cells, monocytes, NK, LAK cells, and on oligodendroglial cells in vitro. In addition, IL-2 plays a role in hematopoiesis, tumor surveillance and anti-inflammatory reactions and hence is a central regulator of the immune response. Non-glycosylated IL-2 is biologically active. Applications Reported: Recombinant Human IL-2 ELISA Lyophilized Standard has been reported for use in the Human IL-2 Ready-Set-Go! ELISA kit (Product # 88-7025). Applications Tested: The Recombinant Human IL-2 ELISA Lyophilized Standard is intended to be used as a standard for the Human IL-2 Ready-Set-Go! ELISA kit (Product # 88-7025-86 . Typically the standard is run in a series of 7 doubling dilutions from the 'top' standard. Please refer to the Certificate of Analysis included with each ELISA set for instructions on the proper reconstitution and dilution of the lyophilized standard. Note: These are single-use vials. Use one time within 4 hours of reconstitution and discard. Reconstitute and dilute as suggested in the Certificate of Analysis or recovery may not be accurate.
Specifications
Specifications
| Accession Number | P60568 |
| For Use With (Application) | Neutralization, ELISA Standard |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 3558 |
| Molecular Weight (g/mol) | 15 kDa |
| Name | Human IL-2 ELISA Standard 10X |
| Purification Method | Purified |
| Quantity | 1 Vial |
| Source | E. Coli |
| Storage Requirements | 4°C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.