Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Human IL-2 QuantiGlo ELISA Kit quantifies human IL-2 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).
Specifications
Specifications
| Assay Range | 1.70 - 1250pg/mL |
| Assay Sensitivity | 0.25pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
| Specificity | Natural and Recombinant Human IL-2 |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3558 |
| Sample Volume | Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
