Learn More
Description
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Description | 0.066pg/mL sensitivity |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
| Type | Human IL-2 Quantikine HS ELISA Kit |
| Gene ID (Entrez) | 3558 (Human); 16183 (Mouse); 116562 (Rat); 396868 (Porcine); 280822 (Bovine); 403989 (Canine); 100034204 (Equine); 751114 (Feline); 100302458 (Rabbit) |
| Includes | Microplate, conjugate, standards, assay diluents, calibrator diluents, wash buffer, color reagent, stop Solution, plate sealers |
| Intended Use | Measure human IL-2 levels in serum and plasma |
| Product Line | R&D Systems |
| Time Range | 4 hours |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
