Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-2 Quantikine QuicKit ELISA
SDP

Catalog No. QK202 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QK202 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. QK202 Supplier R&D Systems™ Supplier No. QK202
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

From the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

  • A one step, 80-minute solid phase ELISA designed to measure human IL-2 levels in cell culture supernates, serum, and plasma.
  • It contains E. coli-expressed recombinant human IL-2 and antibodies raised against the recombinant protein.
  • Results obtained for naturally occurring human IL-2 showed linear curves that were parallel to the standard curves obtained using the recombinant QuicKit™ standards.
  • These results indicate that this kit can be used to determine relative mass values for natural IL-2.

Specifications

Product Type ELISA
Specificity Natural and Recombinant Human IL-2
Gene ID (Entrez) 3558
Host Species Human
Quantity 1 Kit
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym aldesleukin, IL2, IL-2 lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Target IL-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.