Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-20 Quantikine ELISA Kit
SDP

Catalog No. DL200 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DL200 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DL200 Supplier R&D Systems™ Supplier No. DL200

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human IL-20 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-20 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-20 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).

Specifications

Assay Range 62.50 - 4000pg/mL
Assay Sensitivity 16.6pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Human IL-20
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 50604
Sample Volume Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Cytokine Zcyto10, IL10D, IL20, IL-20 ZCYTO10 four alpha helix cytokine, interleukin 20, interleukin-20, MGC96907
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.