Learn More
ACROBiosystems Human IL-21 R 200ug
Supplier:  ACROBiosystems ILRH82F7200UG
Encompass_Preferred
Biotinylated Human IL-21 R Protein FcAvitag (ILR-H82F7) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession # Q9HBE5-1).
Catalog No. 50-253-4573
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.