Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-22 R alpha 1 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2770T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2770T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2770T100 Supplier R&D Systems Supplier No. FAB2770T100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-22R alpha 1 Monoclonal specifically detects IL-22R alpha 1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-22R alpha 1
Applications Flow Cytometry
Classification Monoclonal
Clone 305405
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q8N6P7
Gene Alias CRF2-9, CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R alpha 1, IL22R1, IL22RA1, IL-22RA1, IL-22R-alpha-1, IL-TIF-R1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11
Gene Symbols IL22RA1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human IL-22 R alpha 1 Pro18-Thr228 Accession # Q8N6P7
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58985
Test Specificity Detects human IL-22 R alpha 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-10, rhIL-22BP or rhIL-20 R alpha is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.