Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-22BP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-5316947
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1087SP 25 μg
MAB1087100 100 μg
MAB1087500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1087SP Supplier R&D Systems Supplier No. MAB1087SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-22BP/IL22 RA2 Monoclonal specifically detects IL-22BP/IL22 RA2 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-22BP/IL22 RA2
Applications Western Blot
Classification Monoclonal
Clone 214518
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_851826
Gene Alias class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, CRF2-X, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 RA2, IL-22 receptor subunit alpha-2, IL22BP, IL-22BPCRF2X, IL22RA2, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16
Gene Symbols IL22RA2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-22BP Thr22-Pro231 Accession # NP_851826
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116379
Test Specificity Detects human IL-22BP in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-10, rhIL-22, or rhIL-22 R is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.