Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human IL-23 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP349796 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP349796 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP349796 Supplier Novus Biologicals™ Supplier No. NBP349796
Add to Cart
Add to Cart

Colorimetric Assay Kit

Specifications

Assay Range 12.5 to 800 pg/mL
Assay Sensitivity 7.5 pg/mL
Detection Method Chemiluminescence
Sample Type Serum, Plasma, Other Biological Fluids
Specificity This kit recognizes Human IL-23 in. samples. No significant cross-reactivity or interference between Human IL-23 and analogues was observed
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 51561
Gene Symbol IL23A
Quantity 1 Kit
Research Discipline Cancer, Cytokine Research, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12 B, IL-23, IL-23 subunit alpha, IL-23 A, IL-23-A, IL-23p19, IL23P19P19, interleukin 12, p40, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-12 beta chain, interleukin-12 subunit beta, Interleukin-23 subunit p19, interleukin-six, G-CSF related factor, JKA3 induced upon T-cell activation, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, P19, SGRF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.