Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-27 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2526
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2526 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2526 Supplier R&D Systems Supplier No. AF2526
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-27 Polyclonal specifically detects IL-27 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-27
Applications Western Blot, Neutralization, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Neutralization, ELISA Capture (Matched Antibody Pair) 0.2-0.8 μg/mL
Gene Accession No. Q14213 (EBI-3 subunit), AAM34498 (p28 subunit)
Gene Alias IL27, IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A
Gene Symbols IL27
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-27 (R&D Systems, Catalog # 2526-IL) Arg21-Lys229 (IL-27 EBI-3 subunit), Phe29-Pro243 (IL-27 p28 subunit) Accession # Q14213 (EBI-3 subunit), AAM34498 (p28 subunit)
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246778
Test Specificity Detects human IL-27 in ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.