Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-27 Recombinant Protein, PeproTech®

Catalog No. 200381MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
200-38-1MG 1 mg
20038100UG 100 μg
200-38-10UG 10 μg
200-38-2UG 2 μg
20038500UG 500 μg
200-38-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 200-38-1MG Supplier Gibco™ Supplier No. 200381MG
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-27 produced from HEK293 cells is a non-disulfide-linked, heterodimeric protein composed of a 209 amino acid length EBI3 subunit and a 215 amino acid length IL-27-p28 subunit, for a total sequence length of 424 amino acid residues. The calculated molecular weight of the associated IL-27 subunits is 47.8 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-27 (Interleukin 27) is a cytokine belonging to the IL-12 family, and is a heterodimeric cytokine composed of EBI3 and p28 subunits. IL-27 is produced by activated dendritic cells and macrophages in response to Toll-like receptor ligands and pro-inflammatory cytokines. IL-27 promotes CD4+ T cell differentiation to the Th1 lineage by inducing expression of the transcription factor T-bet and up regulating IL-12R beta2, which in turn suppresses Th2 and Th17 differentiation and proliferation. IL-27 is a recently discovered member of the IL-6/IL-12 family of proinflammatory and immunoregulatory cytokines. IL-27 acts by binding to its receptor WSX-1 and gp130 which results in the activation of a Jak/STAT signaling cascade, suggesting IL-27 involvement in the regulation of immune processes. It has been suggested that IL-27 can also be used as a therapeutic agent against cancer as it can also induce tumor-specific anti-tumor activity mediated through CD8+ T-cells, IFN-gamma, and T-bet. In humans, the gene is located on the p arm of chromosome 16.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14213, Q8NEV9
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 10148, 246778
Molecular Weight (g/mol) 47.8 kDa
Name Human IL-27
Quantity 1 mg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; Il27; IL-27; IL27 p28; IL-27 p28 subunit; IL27 subunit; IL-27 subunit alpha; IL-27 subunit beta; Il27a; IL-27 A; IL-27-A; IL27-A; Il27b; IL-27 B; IL27p28; IL-27p28; Il30; IL35 subunit; ILN; Interleukin; interleukin 27; interleukin 30; interleukin-27 subunit alpha; Interleukin-27 subunit beta; interleukin-30; MGC71873; M-IL-27 (p28); p28; RGD1561420
Common Name IL-27
Gene Symbol Ebi3, IL27
Biological Activity Determined by its ability to stimulate the proliferation of human TF-1 cells. The expected ED50 range is 400-500 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence p28 subunit: FPRPPGRPQL SLQELRREFT VSLHLARKLL SEVRGQAHRF AESHLPGVNL YLLPLGEQLP DVSLTFQAWR RLSDPERLCF ISTTLQPFHA LLGGLGTQGR WTNMERMQLW AMRLDLRDLQ RHLRFQVLAA GFNLPEEEEE EEEEEEEERK GLLPGALGSA LQGPAQVSWP QLLSTYRLLH SLELVLSRAV RELLLLSKAG HSVWPLGFPT LSPQP EBI3 subunit: RKGPPAALTL PRVQCRASRY PIAVDCSWTL PPAPNSTSPV SFIATYRLGM AARGHSWPCL QQTPTSTSCT ITDVQLFSMA PYVLNVTAVH PWGSSSSFVP FITEHIIKPD PPEGVRLSPL AERQLQVQWE PPGSWPFPEI FSLKYWIRYK RQGAARFHRV GPIEATSFIL RAVRPRARYY VQVAAQDLTD YGELSDWSLP ATATMSLGK
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.