Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-28B/IFN-lambda 3 Quantikine ELISA Kit
SDP

Catalog No. D28B00 Shop All R&D Systems Products
Change view
Click to view available options
Sample Type:
Cell Culture Supernatant, Cell Lysates, Plasma (EDTA), Serum, Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
D28B00 Cell Culture Supernatant, Cell Lysates, Plasma (EDTA), Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. D28B00 Supplier R&D Systems™ Supplier No. D28B00
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the quantitative determination of human Interleukin 28B (IL-28B) concentrations in cell culture supernates, cell lysates, serum, EDTA plasma, and urine. The Quantikine Human IL-28B/IFN-lambda 3 Immunoassay is a 4.5 hour solid phase ELISA.

The Human IL-28B/IFN-lambda 3 Quantikine ELISA Kit quantifies human IL-28B/IFN-lambda 3 in cell culture supernates (50 ul), cell lysates (100 ul), serum (100 ul), edta plasma (100 ul), urine (50 ul).

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 1.87pg/mL
Conjugate HRP
Cross Reactivity <0.5% cross-reactivity observed with available related molecules. <50% cross-species reactivity observed with species tested.
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA), Serum, Urine
Specificity Natural and Recombinant Human IL-28 B.
For Use With (Application) ELISA
Gene ID (Entrez) 282617 (Human); 338374 (Mouse)
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (100uL),Serum (100uL),EDTA Plasma (100uL),Urine (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym interleukin-28 B, IFN-lambda 3, IL28B, IL-28 B, IL28C, interferon, lambda 3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.