Learn More
FUJIFILM IRVINE SCIENTIFIC INC Human IL-3 , Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 100801MG
Encompass_Preferred
Interleukin 3 (IL-3) is a cytokine that is produced by activated T cells and mast cells. IL-3 induces the differentiation of hematopoietic stem cells into myeloid precursor cells, such as erythrocyte, megakaryocyte, granulocyte, monocyte, and dendritic cells. IL-3 also functions in the nervous system and is important during the B-1 cell regulation of chronic inflammatory diseases.
Catalog No. 50-900-472
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.