Learn More
ACROBiosystems ACROBiosystems Human IL-3 R alpha / CD123 Protein, Fc Tag (MALS & SPR verified)
Supplier:  ACROBiosystems ILAH5252100UG
Encompass_Preferred
Recombinant Protein;100UG;Human IL-3 R alpha, Fc Tag (ILA-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305 (Accession # P26951-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 59.5 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Anti-IL-3 R alpha Antibody, Human IgG1 immobilized on CM5 Chip can bind Human IL-3 R alpha, Fc Tag (Cat. No. ILA-H5252) with an affinity constant of 50.3 nM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-210-6241