Learn More
ACROBiosystems Human IL-31 RA 25ug
Supplier:  ACROBiosystems ILAH82F625UG
Encompass_Preferred
Biotinylated Human IL-31 RA Protein FcAvitag (ILA-H82F6) is expressed from human 293 cells (HEK293). It contains AA Ala 20 – Glu 519 (Accession # Q8NI17-1).
Catalog No. 50-253-4597
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.