Learn More
Bon Opus Biosciences Recombinant Human IL-33 - Source: E.coli
SDP
Supplier:  Bon Opus Biosciences BP22650UG
Encompass_Preferred
Recombinant Human IL-33 is produced by our E.coli expression system and the target gene encoding Ser112-Thr270 is expressed. Accession#O95760. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4
Catalog No. NC2733665
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.