Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-36 alpha (IL-1F6) Recombinant Protein, PeproTech®

Catalog No. 20036A250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 μg
2 x 500 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20036A250UG 250 μg
20036A100UG 100 μg
20-036-A01M 2 x 500 μg
200-36A-2UG 2 μg
20036A500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 20036A250UG Supplier Gibco™ Supplier No. 20036A250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

200-36A-1MG will be provided as 2 x 500 μg (200-36A-500UG). Recombinant Human IL-36alpha is a 17.2 kDa protein containing 154 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The IL-1 family is comprised of 11 structurally related ligands, including recently re-named IL-36alpha (IL-1F6), beta (IL-1F8), and gamma (IL-1F9). IL-36alpha is highly expressed in psoriatic plaques, as are IL-36beta and IL-36gamma, and is the only IL-1 family member found to be expressed by T cells, as well as monocytes and B cells. Primarily found in skin and lymphoid tissues, IL-36alpha is also found to a lesser extent in tissues of the fetal brain, trachea, stomach, and intestine. IL-36alpha signals through a combination of the IL-1Rrp2 (IL-1R6) receptor, which is primarily expressed on certain dendritic cells, and the widely expressed IL-RAcP (IL-1 receptor accessory protein). IL-36alpha also functions as an agonist of NF-kappaB and MAPK pathways, and induces the production of proinflammatory cytokines and chemokines, including IL-6, IL-8, BD-2, and BD-3.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UHA7
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 27179
Molecular Weight (g/mol) 17.2 kDa
Name Human IL-36 alpha (IL-1F6)
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias FIL1; FIL1 epsilon; FIL1(EPSILON); Fil1e; Fil1epsilon; IL-1 epsilon; IL1(EPSILON); Il1e; Il1f6; IL-1F6; IL-1F6 (FIL-1-epsilon); Il1f9; Il1h1; IL-1H1; IL1RP2; Il36a; Interleukin; interleukin 1; interleukin 1 family, member 6; interleukin 1 family, member 6 (epsilon); interleukin 1 family, member 9; interleukin 1, epsilon; interleukin 36, alpha; interleukin-1 epsilon; Interleukin-1 family member 6; interleukin-1 homolog 1; Interleukin-36; Interleukin-36 alpha
Common Name IL1F6
Gene Symbol IL36A
Biological Activity Measured by its ability to induce IL-8 secretion by A431 human epidermoid carcinoma cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MKIDTPQQGS IQDINHRVWV LQDQTLIAVP RKDRMSPVTI ALISCRHVET LEKDRGNPIY LGLNGLNLCL MCAKVGDQPT LQLKEKDIMD LYNQPEPVKS FLFYHSQSGR NSTFESVAFP GWFIAVSSEG GCPLILTQEL GKANTTDFGL TMLF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.