Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-36 gamma (IL-1F9), Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF20036G1MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF20036G1MG 1 mg
AF20036G10 10 μg
AF20036G100 100 μg
AF20036G500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AF20036G1MG Supplier Gibco™ Supplier No. AF20036G1MG
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-36gamma is a 17.0 kDa protein containing 152 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Cytokine that binds to and signals through the IL1RL2/IL-36R receptor which in turn activates NF-kappa-B and MAPK signaling pathways in target cells. Part of the IL-36 signaling system that is thought to be present in epithelial barriers and to take part in local inflammatory response; similar to the IL-1 system with which it shares the coreceptor IL1RAP. Seems to be involved in skin inflammatory response by acting on keratinocytes, dendritic cells and indirectly on T-cells to drive tissue infiltration, cell maturation and cell proliferation. In cultured keratinocytes induces the expression of macrophage, T-cell, and neutrophil chemokines, such as CCL3, CCL4, CCL5, CCL2, CCL17, CCL22, CL20, CCL5, CCL2, CCL17, CCL22, CXCL8, CCL20 and CXCL1; also stimulates its own expression and that of the prototypic cutaneous proinflammatory parameters TNF-alpha, S100A7/psoriasin and inducible NOS. May play a role in proinflammatory responses during particular neutrophilic airway inflammation: activates mitogen-activated protein kinases and NF-kappa B in primary lung fibroblasts, and stimulates the expression of IL-8 and CXCL3 and Th17 chemokine CCL20 in lung fibroblasts. May be involved in the innate immune response to fungal pathogens, such as Aspergillus fumigatus.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NZH8
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 56300
Molecular Weight (g/mol) 17 kDa
Name Human IL-36 gamma (IL-1F9), Animal-Free
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias IL-1 epsilon; IL-1 related protein 2; IL-1(EPSILON); IL1E; IL-1-epsilon; IL1F9; IL-1F9; IL1H1; IL-1H1; IL-1-related protein 2; IL1RP2; IL-1 RP2; IL36G; Interleukin; interleukin 1 family, member 9; interleukin 1-related protein 2; interleukin 36, gamma; interleukin-1 epsilon; Interleukin-1 family member 9; interleukin-1 homolog 1; Interleukin-36; Interleukin-36 gamma; UNQ2456/PRO5737
Common Name IL1F9
Gene Symbol IL36G
Biological Activity Measured by its ability to induce IL-8 production by human PBMCs.
Conjugate Unconjugated
Recombinant Recombinant
Sequence SMCKPITGTI NDLNQQVWTL QGQNLVAVPR SDSVTPVTVA VITCKYPEAL EQGRGDPIYL GIQNPEMCLY CEKVGEQPTL QLKEQKIMDL YGQPEPVKPF LFYRAKTGRT STLESVAFPD WFIASSKRDQ PIILTSELGK SYNTAFELNI ND
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.