Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Human IL-36 gamma/IL-1F9 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-36 gamma/IL-1F9.
Specifications
Specifications
| Assay Range | 18.80 to 1,200pg/mL |
| Immunoassay Kit Format | 96-well Plate |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
| Sufficient For | Five 96-well plates |
| Target Species | Human |
| Gene ID (Entrez) | 56300 (Human); 215257 (Mouse); 499744 (Rat) |
| Kit Contents | 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Target | Interleukin 36 gamma/Interleukin 1 Family 9 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
