Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-36RA Recombinant Protein, PeproTech®

Catalog No. 20036RA250
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20036RA250 250 μg
20036RA100 100 μg
20036RA1MG 1 mg
20036RA25UG 25 μg
20036RA500 500 μg
20036RA5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 20036RA250 Supplier Gibco™ Supplier No. 20036RA250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-36RA is an E.coli-derived 17 kDa protein containing 154 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine was shown to specifically inhibit the activation of NF-kappaB induced by interleukin 1 family, member 6 (IL1F6). This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Two alternatively spliced transcript variants encoding the same protein have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UBH0
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 26525
Molecular Weight (g/mol) 17 kDa
Name Human IL-36 RA
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI413231; FIL1; FIL1 delta; FIL1(DELTA); FIL1D; Fil1delta; IL-1 delta; IL-1 related protein 3; Il1f5; IL-1F5; IL1F5 (Canonical product IL-1F5a); IL-1F5 (IL-1 HY1, FIL1-delta, IL-1 RP3, IL-1L1, IL-1-delta); Il1h3; Il-1h3; IL1HY1; IL-1 HY1; IL1L1; IL-1L1; IL-1 ra homolog 1; IL-1-related protein 3; IL1RP3; IL-1 RP3; IL36RA; IL-36 Ra; IL36RN; Interleukin; interleukin 1 family, member 5; interleukin 1 family, member 5 (delta); interleukin 1 receptor antagonist homolog 1; interleukin 36 receptor antagonist; interleukin-1 delta; Interleukin-1 family member 5; interleukin-1 family member 5; interleukin-36 receptor antagonist protein; interleukin-1 homolog 3; Interleukin-1 HY1; interleukin-1 receptor antagonist homolog 1; Interleukin-1-like protein 1; interleukin-36 receptor antagonist protein; MGC29840; PSORP; PSORS14; UNQ1896/PRO4342
Common Name IL1F5
Gene Symbol IL36RN
Biological Activity Measured by its ability to inhibit secretion of IL-8 by A431 cells in the presence of IL-36 gamma.
Conjugate Unconjugated
Recombinant Recombinant
Sequence VLSGALCFRM KDSALKVLYL HNNQLLAGGL HAGKVIKGEE ISVVPNRWLD ASLSPVILGV QGGSQCLSCG VGQEPTLTLE PVNIMELYLG AKESKSFTFY RRDMGLTSSF ESAAYPGWFL CTVPEADQPV RLTQLPENGG WNAPITDFYF QQCD
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.