Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-37 (IL-1F7) Recombinant Protein, PeproTech®

Catalog No. 200391MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
200-39-1MG 1 mg
20039100UG 100 μg
20039250UG 250 μg
200-39-25UG 25 μg
20039500UG 500 μg
200-39-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 200-39-1MG Supplier Gibco™ Supplier No. 200391MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-37 is a 19.4 kDa protein consisting of 174 amino acids, corresponding to the mature activated form of IL-37. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine can bind to, and may be a ligand for interleukin 18 receptor (IL18R1/IL-1Rrp). This cytokine also binds to interleukin 18 binding protein (IL18BP), an inhibitory binding protein of interleukin 18 (IL18), and subsequently forms a complex with IL18 receptor beta subunit, and through which it inhibits the activity of IL18. This gene along with eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Five alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NZH6
For Use With (Application) ELISA, Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 27178
Molecular Weight (g/mol) 19.4 kDa
Name Human IL-37 (IL-1F7)
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias FIL1; FIL1 zeta; FIL1(ZETA); FIL1Z; IL-1 zeta; IL1F7; IL-1F7; IL1F7 (canonical product IL-1F7b); IL-1F7b (IL-1H4, IL-1 H, IL-1 RP1); IL-1 H; IL1H4; IL-1H4; IL1RP1; IL-1 RP1; IL-1 X; IL-1 x protein; IL37; IL-37; ILN; Interleukin; interleukin 1 family member 7; interleukin 1, zeta; interleukin 37; Interleukin-1 family member 7; Interleukin-1 homolog 4; interleukin-1 superfamily z; Interleukin-1 zeta; Interleukin-1-related protein; interleukin-23; Interleukin37; Interleukin-37
Common Name IL-37
Gene Symbol IL37
Biological Activity Measured by its ability to bind Interleukin-18 Binding Protein isoform a (IL-18 BPa) in functional ELISA.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MVHTSPKVKN LNPKKFSIHD QDHKVLVLDS GNLIAVPDKN YIRPEIFFAL ASSLSSASAE KGSPILLGVS KGEFCLYCDK DKGQSHPSLQ LKKEKLMKLA AQKESARRPF IFYRAQVGSW NMLESAAHPG WFICTSCNCN EPVGVTDKFE NRKHIEFSFQ PVCKAEMSPS EVSD
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.