Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-4 Receptor alpha (soluble) Recombinant Protein, PeproTech®

Catalog No. 20004RC5UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20004RC5UG 5 μg
20004RC100 100 μg
20004RC1MG 1 mg
20004RC20UG 20 μg
20004RC250 250 μg
20004RC500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 20004RC5UG Supplier Gibco™ Supplier No. 20004RC5UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human sIL-4 Receptor alpha is a 23.9 kDa glycoprotein corresponding to 209 amino acid residues of the extracellular domain of IL-4Ralpha. As a result of glycosylation, Recombinant Human sIL-4 Receptor alpha migrates with an apparent molecular mass of approximately 50-65 kDa by SDS-PAGE gel, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL4 Receptor alpha (IL4RA) is the alpha chain of the interleukin 4 receptor which binds to both interleukin 4 and interleukin 13 to regulate IgE production, chemokine and mucus production at sites of allergic inflammmation. The IL4 response is involved in promoting Th2 differentiation. The secreted extracellular domain of IL4R alpha, called sIL4R alpha, can inhibit IL4-mediated cell proliferation and IL5 upregulation by T-cells. It has no signaling abilities. Allelic variations in the IL4RA gene have been associated with atopy, a condition that can manifest itself as allergic rhinitis, sinusitus, asthma, or eczema.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P24394
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 3566
Molecular Weight (g/mol) 23.9 kDa
Name Human IL-4 Receptor alpha (soluble)
Quantity 5 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 582J2.1; CD124; il-4 receptor; IL-4 receptor alpha chain; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; Il4r; IL4R nirs variant 1; IL-4 R subunit alpha; IL4RA; IL-4 RA; IL-4 R-alpha; interleukin 4 receptor; interleukin 4 receptor, alpha; interleukin-4 receptor alpha chain; interleukin-4 receptor subunit alpha; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha
Common Name IL4R
Gene Symbol IL4R
Biological Activity The ED50 was determined by its ability to inhibit the IL-4 dependent proliferation of human TF-1 cells is ≤ 5.0 ng/ml (in the presence of 0.5 ng/ml of IL-4), corresponding to a specific activity of ≥ 2 x 10^5 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence GNMKVLQEPT CVSDYMSIST CEWKMNGPTN CSTELRLLYQ LVFLLSEAHT CIPENNGGAG CVCHLLMDDV VSADNYTLDL WAGQQLLWKG SFKPSEHVKP RAPGNLTVHT NVSDTLLLTW SNPYPPDNYL YNHLTYAVNI WSENDPADFR IYNVTYLEPS LRIAASTLKS GISYRARVRA WAQCYNTTWS EWSPSTKWHN SYREPFEQH
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.