Learn More
ACROBiosystems Human IL-5 R alpha, Fc Tag (ILA-H5269) is expressed from human 293 cells (HEK293). It contains AA Asp 21 - Trp 342 (Accession # Q01344-1).
Supplier:  ACROBiosystems ILAH5269100UG
Encompass_Preferred
Interleukin-5 receptor subunit alpha (IL5RA) is also known as IL-5 receptor subunit alpha, IL-5R subunit alpha and CD125,which is a single-pass type I membrane protein that belongs to the type I cytokine receptor family and type 5 subfamily.IL5R, a heterodimer of an alpha and a beta subunit, is expressed on eosinophils and basophils. IL5RA / CD125 is the receptor for interleukin-5 (IL5) and the alpha chain can bind to IL5. The WSXWS motif of IL5RA appears to be necessary for proper protein folding and thereby efficient intracellular transport and cell-surface receptor binding.The beta subunit of IL5R is common to the IL3, IL5 and GM-CSF receptors.
Catalog No. 50-223-0965
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.