Learn More
ACROBiosystems ACROBiosystems Human IL-5 R alpha / CD125 Protein, His Tag
Supplier:  ACROBiosystems ILAH52H41MG
Encompass_Preferred
Recombinant Protein;1MG;Human IL-5 R alpha, His Tag (ILA-H52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 21 - Glu 335 (Accession # NP_000555).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.7 kDa. The protein migrates as 48-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6677
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.