Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-5 Recombinant Protein, PeproTech®

Catalog No. 20005100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20005100UG 100 μg
200-05-10UG 10 μg
200-05-1MG 1 mg
20005250UG 250 μg
200-05-2UG 2 μg
20005500UG 500 μg
200-05-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 20005100UG Supplier Gibco™ Supplier No. 20005100UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-5 is a 26.5 kDa disulfide-linked homodimeric protein containing two 116 amino acid chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05113
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 3567
Molecular Weight (g/mol) 26.5 kDa
Name Human IL-5
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias B-cell differentiation factor I; B-cell growth factor II; BCGF-II; colony-stimulating factor, eosinophil; cytotoxic T-lymphocyte inducer; EDF; Eosinophil differentiation factor; IL5; IL-5; ILN; Interleukin; Interleukin 5; Interleukin 5 (colony-stimulating factor eosinophil); Interleukin 5 (colony-stimulating factor, eosinophil); Interleukin5; interleukin-5; T-cell replacing factor; TRF
Common Name IL-5
Gene Symbol Il5
Biological Activity The ED50 as determined by the dose-dependent proliferation of TF-1 cells was ≤ 0.15 ng/ml, corresponding to a specific activity of ≥ 6.6 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence MIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIES
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.