Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-6 QuantiGlo ELISA Kit
SDP

Catalog No. SQ6000B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SQ6000B 1 Pk. (6 Plates)
Q6000B 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SQ6000B Supplier R&D Systems™ Supplier No. SQ6000B
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human IL-6 QuantiGlo ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-6 QuantiGlo ELISA Kit quantifies human IL-6 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).

Specifications

Assay Range 0.48 to 1500 pg/mL
Assay Sensitivity 0.35 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Citrate, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human IL-6
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3569
Gene Symbol IL6
Quantity 1 Pk. (6 Plates)
Sample Volume Cell Culture Supernates (100 μL), Serum (100 μL), EDTA Plasma (100 μL), Heparin Plasma (100 μL), Citrate Plasma (100 μL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2 A
Target IL-6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.