Learn More
Description
Specifications
Specifications
| Assay Range | 3.13 - 300 pg/mL |
| Assay Sensitivity | 0.626 pg/mL |
| Sample Type | Cell Culture Supernatant, Serum, Plasma (EDTA), Plasma (Heparin), Urine |
| Specificity | Natural and recombinant human IL-6. |
| Gene ID (Entrez) | 3569 |
| Quantity | 1 Pack (6 Plates) |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
