Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-6 Quantikine HS ELISA
SDP

Catalog No. SS600C Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Pk. (50 Plates)
1 Pk. (6 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SS600C 1 Pk. (6 Plates)
PHS600C 1 Pk. (50 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SS600C Supplier R&D Systems™ Supplier No. SS600C

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human IL-6 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-6 Quantikine HS ELISA Kit quantifies human IL-6 in serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), urine (100 ul).

1 Pack (50 Plates)

Specifications

Assay Range 0.16 to 10 pg/mL
Assay Sensitivity 0.09 pg/mL
Product Line R&D Systems
Sample Type Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human IL-6
For Use With (Application) ELISA
Gene ID (Entrez) 3569
Host Species Human
Quantity 1 Pk. (6 Plates)
Sample Volume Serum (100 uL),EDTA Plasma (100 uL),Heparin Plasma (100 uL),Urine (100 uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2 A
Target IL-6
Total Assay Time 4 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.