Learn More
Gibco™ Human IL-6 Recombinant Protein, PeproTech®
Description
Recombinant Human IL-6 is a 20.9 kDa protein containing 184 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P05231 |
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 3569 |
| Molecular Weight (g/mol) | 20.9 kDa |
| Name | Human IL-6 |
| Quantity | 20 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.